Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse Wnt-8a Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2248
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2248 100 μg
AF2248SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF2248 Supplier R&D Systems Supplier No. AF2248
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Wnt-8a Polyclonal specifically detects Wnt-8a in Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Wnt-8a
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. Q64527
Gene Alias protein Wnt-8a, Protein Wnt-8d, wingless-type MMTV integration site family, member 8A, Wnt8a, WNT8DWNT8d
Gene Symbols WNT8A
Host Species Goat
Immunogen E. coli-derived recombinant mouse Wnt-8a peptide Trp23-Glu53, Leu196-Arg273 Accession # Q64527
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7478
Test Specificity Detects mouse Wnt-8a in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) Wnt-1, rmWnt-3A, rmWnt-4, rmWnt-5A, and rmWnt-10B is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.