Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse Wnt-9b Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Supplier: R&D Systems AF3669
This item is not returnable.
View return policy
Description
Wnt-9b Polyclonal specifically detects Wnt-9b in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Wnt-9b | |
Polyclonal | |
Unconjugated | |
O35468 | |
WNT9B | |
E. coli-derived recombinant mouse Wnt-9b Ala46-Glu90, Lys228-Tyr298 Accession # O35468 | |
100 μg | |
Primary | |
Detects mouse Wnt-9b in direct ELISAs and Western blots. In Western blots, less than 5% cross-reactivity with recombinant human (rh) Wnt-2, rhWnt-9a, recombinant mouse (rm) Wnt-1, rmWnt-2b, rmWnt-3a, rmWnt-4, rmWnt-5a, rmWnt-5b, rmWnt-11, and rmWnt-16 is observed. | |
Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
Protein Wnt-14b, wingless-type MMTV integration site family, member 15, wingless-type MMTV integration site family, member 9B, Wnt-14b, WNT14Bprotein Wnt-9b, Wnt-15, WNT15Protein Wnt-15, Wnt9b | |
Goat | |
Affinity Purified | |
RUO | |
7484 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction