Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse XCL1/Lymphotactin Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB486SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB486SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB486SP Supplier R&D Systems Supplier No. MAB486SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

XCL1/Lymphotactin Monoclonal specifically detects XCL1/Lymphotactin in Mouse samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).

Specifications

Antigen XCL1/Lymphotactin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 80222
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. AAA56752
Gene Alias ATAC, ATACSmall-inducible cytokine C1, C motif chemokine 1, chemokine (C motif) ligand 1, Cytokine SCM-1, LPTNSCM-1-alpha, LTNXC chemokine ligand 1, lymphotactin, lymphotaxin, SCM-1, SCM-1 alpha, SCM-1a, SCM1A, SCYC1SCM1, single cysteine motif 1a, small inducible cytokine subfamily C, member 1 (lymphotactin)
Gene Symbols XCL1
Host Species Rat
Immunogen E. coli-derived recombinant mouse XCL1/Lymphotactin Val22-Gly114 Accession # AAA56752
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6375
Test Specificity Detects mouse XCL1/Lymphotactin in ELISAs and Western blots. In sandwich immunoassays, less than 1% cross-reactivity with recombinant human XCL1/Lymphotactin is observed and no cross-reactivity with recombinant viral (rv) CMV UL147 is observed. In Western blots, 75% cross-reactivity with rvCMV UL147 is observed and no cross-reactivity with recombinant human CCL1-3, 5, 7, 8, 11, 13-25, 28, recombinant mouse CCL1-7, 9, 11, 12, 15, 19, 21, 22, 24, 25, 27 or 28 is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.