Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mouse ZIP4H/Tex11 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5627SP
This item is not returnable.
View return policy
Description
ZIP4H/Tex11 Polyclonal specifically detects ZIP4H/Tex11 in Mouse samples. It is validated for Western Blot.Specifications
ZIP4H/Tex11 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
testis expressed 11, testis expressed sequence 11, testis-expressed sequence 11 protein, Tex11, TGC1, TSGA3 | |
Goat | |
Affinity Purified | |
RUO | |
56159 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q14AT2 | |
TEX11 | |
E. coli-derived recombinant mouse ZIP4H/Tex11 Met768-Leu947 Accession # Q14AT2 | |
25 μg | |
Primary | |
Detects mouse ZIP4H/Tex11 in Western blots. | |
Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction