Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse ZNRF3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB8328SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8328SP 25 μg
MAB8328100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB8328SP Supplier R&D Systems Supplier No. MAB8328SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

ZNRF3 Monoclonal specifically detects ZNRF3 in Mouse samples. It is validated for Immunocytochemistry.

Specifications

Antigen ZNRF3
Applications Immunocytochemistry
Classification Monoclonal
Clone 2035A
Conjugate Unconjugated
Dilution Immunocytochemistry 5-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. Q5SSZ7
Gene Alias BK747E2.3, KIAA1133, novel C3HC4 type Zinc finger (ring finger), RNF203, zinc and ring finger 3, zinc/RING finger protein 3
Host Species Rabbit
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse ZNRF3 Lys53-Met216 Accession # Q5SSZ7
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 84133
Test Specificity Detects mouse ZNRF3 in direct ELISAs.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.