Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOV10 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10077314C
Description
MOV10 Polyclonal antibody specifically detects MOV10 in Human, Mouse samples. It is validated for Western BlotSpecifications
MOV10 | |
Polyclonal | |
Western Blot | |
DKFZp667O1423, EC 3.6.1, EC 3.6.4.13, FLJ32791, KIAA1631mouse) homolog, Mov10, Moloney leukemia virus 10, homolog (mouse) | |
The immunogen recognized by this antibody maps to a region between residue 953 and 1003 of human Moloney leukemia virus 10, homolog using the numbering given in entry NP_066014.1 (GeneID 4343). | |
0.1 mL | |
Primary | |
Human, Mouse | |
Purified |
Western Blot | |
DyLight 650 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
4343 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction