Learn More
MOV10 Mouse anti-Human, Mouse, Clone: 15C1B8, Bethyl Laboratories
Mouse Monoclonal Antibody
Supplier: Bethyl Laboratories, Inc A500009A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Dog, Horse, Rabbit, Guinea pig_10141, Panda, Orangutan, Monkey, Gorilla, Chimpanzee and Naked mole rat
Probable RNA helicase. Required for RNA-mediated gene silencing by the RNA-induced silencing complex (RISC). Required for both miRNA-mediated translational repression and miRNA- mediated cleavage of complementary mRNAs by RISC. Also required for RNA-directed transcription and replication of the human hepatitis delta virus (HDV). Interacts with small capped HDV RNAs derived from genomic hairpin structures that mark the initiation sites of RNA-dependent HDV RNA transcription.Specifications
MOV10 | |
Monoclonal | |
Unconjugated | |
MOV10 | |
Putative helicase MOV-10, Moloney leukemia virus 10 protein, MOV10, KIAA1631 | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
17454, 4343 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
15C1B8 | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P23249, Q9HCE1 | |
MOV10 | |
Between 953 and 1003 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.