Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOV10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565103
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565103 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565103 Supplier Invitrogen™ Supplier No. PA565103
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEELQMEHDIR HYDLESVPMT WDPVDQNPRL LTLEVPGVTE SRPSVLRGDH LFALLSSETH QEDPITYKGF VHKVELDRVK LSFSMSLLSR FVDGLTFKVN FTFNRQPLRV QHRALELTGR WLLWPMLFPV AP Highest antigen sequence identity to the following orthologs - mouse 94%, rat 95%.

5' to 3' RNA helicase contributing to UPF1 mRNA target degradation by translocation along 3' UTRs (PubMed:24726324). Required for microRNA (miRNA)-mediated gene silencing by the RNA-induced silencing complex (RISC). Required for both miRNA-mediated translational repression and miRNA-mediated cleavage of complementary mRNAs by RISC (PubMed:16289642, PubMed:17507929, PubMed:22791714). In cooperation with FMR1, regulates miRNA-mediated translational repression by AGO2 (PubMed:25464849). Restricts retrotransposition of long interspersed element-1 (LINE-1) in cooperation with TUT4 and TUT7 counteracting the RNA chaperonne activity of L1RE1 (PubMed:30122351, PubMed:23093941). Facilitates LINE-1 uridylation by TUT4 and TUT7 (PubMed:30122351). Required for embryonic viability and for normal central nervous system development and function. Plays two critical roles in early brain development: suppresses retroelements in the nucleus by directly inhibiting cDNA synthesis, while regulates cytoskeletal mRNAs to influence neurite outgrowth in the cytosol. May function as a messenger ribonucleoprotein (mRNP) clearance factor (PubMed:24726324). Exhibits antiviral activity against dengue virus (DENV) (PubMed:27974568). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOV10
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MOV10
Gene Accession No. Q9HCE1
Gene Alias Armitage homolog; C77703; capping protein (actin filament) muscle Z-line, alpha 1; Capza1; fSAP113; functional spliceosome-associated protein 113; gb110; Helicase MOV-10; KIAA1631; Moloney leukemia virus 10; moloney leukemia virus 10 protein; Mov10; Mov-10; Mov10 RISC complex RNA helicase; Mov10, Moloney leukemia virus 10; Mov10, Moloney leukemia virus 10, homolog; putative helicase MOV-10
Gene Symbols MOV10
Host Species Rabbit
Immunogen Recombinant Human MOV10. Recombinant protein control fragment (Product #RP-105859).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4343
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.