Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ MOV10L1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106711
Description
Antibody detects endogenous levels of total MOV10L1.
Isoform 1: ATP-dependent RNA helicase required during spermatogenesis to repress transposable elements and prevent their mobilization, which is essential for germline integrity (PubMed:20534472, PubMed:20547853, PubMed:23166510, PubMed:25762440). Acts via the piRNA metabolic process, which mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and governs the methylation and subsequent repression of transposons (PubMed:20534472, PubMed:20547853, PubMed:23166510, PubMed:25762440). Involved in the primary piRNA metabolic process (PubMed:20534472, PubMed:20547853, PubMed:23166510, PubMed:25762440). Specifically binds to piRNA precursors and promotes the generation of intermediate piRNA processing fragments that are subsequently loaded to Piwi proteins (PubMed:25762440). Acts via its ATP-dependent RNA helicase activity: displays 5'-3' RNA unwinding activity and probably mediates unwinding and funneling of single-stranded piRNA precursor transcripts to the endonuclease that catalyzes the first cleavage step of piRNA processing to generate piRNA intermediate fragments that are subsequently loaded to Piwi proteins (PubMed:25762440). {ECO:0000269; PubMed:20534472, ECO:0000269; PubMed:20547853, ECO:0000269; PubMed:23166510, ECO:0000269; PubMed:25762440}.; Isoform 2: May act downstream of MEF2C during heart formation. Acts as a cardiac-specific suppressor of cardiomyocyte hypertrophy and cell cycle progression, suggesting that it may suppress these processes through the regulation of CDKN1A. Such results however require additional evidence. {ECO:0000305; PubMed:11854500}.
Specifications
MOV10L1 | |
Polyclonal | |
Unconjugated | |
MOV10L1 | |
Cardiac helicase activated by MEF2 protein; cardiac helicase activated by MEF2C protein; cardiac-specific RNA helicase; Champ; Csm; DJ402G11.8; Moloney leukemia virus 10-like 1; moloney leukemia virus 10-like protein 1; Moloney leukemia virus 10-like protein 1 homolog; Mov10 RISC complex RNA helicase like 1; Mov10l1; Mov10l1, Moloney leukemia virus 10-like 1, homolog; Mov10-like 1; MOV10-like protein 1; MOV10-like protein 1 homolog; putative helicase Mov10l1; RNA helicase Mov10l1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
300141, 54456, 83456 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q99MV5, Q9BXT6 | |
MOV10L1 | |
A synthesized peptide derived from human MOV10L1(Accession Q9BXT6), corresponding to amino acid residues Q317-L367. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction