Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MOX1 Antibody (OTI5B11), Alexa Fluor™ 532, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272770AF532
Description
MOX1 Monoclonal specifically detects MOX1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
MOX1 | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
MEOX1 | |
Full length recombinant protein of human MOX1 (NP_004518) produced in E.coli. | |
0.1 mL | |
4222.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI5B11 | |
Western Blot, Immunohistochemistry | |
homeobox protein MOX-1, mesenchyme homeobox 1MOX1mesenchyme homeo box 1 | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction