Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583765
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583765 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583765 Supplier Invitrogen™ Supplier No. PA583765
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WGCLRNPHSE GNGASGLPHY PPTPFSFHQK PDFLATATAA YPDFSASCLA ATPHSLPQEE HIFTEQHPAF PQSPNWHFPV SDARRRPNSG PAGGSKEMGT SSLGLVDTTG GPGDDYGVLG STANETEKKS SR Highest antigen sequence indentity to the following orthologs: Mouse - 74%, Rat - 74%.

MEOX1 belongs to a family of nonclustered, diverged homeobox genes. It may play a role in regulating growth and differentiation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOX1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene MEOX1
Gene Accession No. P50221
Gene Alias AI385561; D330041M02Rik; GP91-2; homeobox protein MOX-1; KFS2; MEOX1; Mesenchyme homeobox 1; mitogenic oxidase 1; MOX1; Mox-1; NOH1; NOH-1; NOX-1
Gene Symbols MEOX1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human MEOX1. Recombinant protein control fragment (Product #RP-100669).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4222
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.