Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOXD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596295
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596295 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596295 Supplier Invitrogen™ Supplier No. PA596295
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AHLAGRGIRL RHFRKGKEMK LLAYDDDFDF NFQEFQYLKE EQTILPGDNL ITECRYNTKD RAEMTWGGLS TRSEMCLSYL LYYPRINLTR CASIPDIMEQ LQFIGVKEIY RPVTTWPFII KSPKQYKNLS FMDAMNKFKW TKKEGLSFNK LVLSLPVNVR CSKTDNAEWS IQGMTALPPD IERPYKAEPL VCGTSSSSSL H; Positive Samples: U-87MG, H460, A-431, Jurkat, Raji, Mouse uterus, Rat brain; Cellular Location: Endoplasmic reticulum membrane, Single-pass type I membrane protein.

The MOXD1 (Monooxygenase, DBH-like 1) gene encodes a protein that is part of the copper type II, ascorbate-dependent monooxygenase family, which includes enzymes such as dopamine beta-hydroxylase. Located on chromosome 6, the MOXD1 protein plays a role in the biosynthesis of bioactive amines, though its specific substrate and biological functions are less well characterized compared to its family members. This enzyme's activity is implicated in various physiological processes, including the metabolism of neurotransmitters and regulation of blood pressure. Research has suggested that MOXD1 might be involved in oncogenesis, with altered expression observed in certain cancer types. Further studies are needed to elucidate its precise physiological roles and potential as a therapeutic target.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOXD1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.36 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene MOXD1
Gene Accession No. Q6UVY6, Q9CXI3
Gene Alias 3230402N08Rik; DBH-like monooxygenase protein 1; Dbhr; DBH-related protein; dJ248E1.1; dopamine-oxygenase; MNCb-5203; monooxygenase DBH like 1; monooxygenase X; monooxygenase, DBH-like 1; MOX; MOXD1; PRO5780; UNQ2493/PRO5780
Gene Symbols MOXD1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 390-590 of human MOXD1 (NP_0563442).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26002, 294119, 59012
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.