Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MOXD1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501984136
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-4136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-4136 Supplier Bioss Supplier No. BS17733R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifications

Antigen MOXD1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene MOXD1
Gene Accession No. Q6UVY6
Gene Alias 3230402N08Rik; DBH-like monooxygenase protein 1; Dbhr; DBH-related protein; dJ248E1.1; dopamine-oxygenase; MNCb-5203; monooxygenase DBH like 1; Monooxygenase X; monooxygenase, DBH-like 1; MOX; MOXD1; PRO5780; UNQ2493/PRO5780
Gene Symbols MOXD1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human MOXD1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26002
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.