Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566566
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566566 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566566 Supplier Invitrogen™ Supplier No. PA566566
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SSAPQEQYGEC GEKSEATQEQ YTESEEQLVA SEEQPSQDGK PDLPKRRLSE GVEPWRGQLK KSPEALKCRL TEGSE Highest antigen sequence identity to the following orthologs - mouse 67%, rat 61%.

Histone acetyltransferase that acetylates lysine residues in histone H3 and histone H4 (in vitro). Component of the MOZ/MORF complex which has a histone H3 acetyltransferase activity. May act as a transcriptional coactivator for RUNX1 and RUNX2. Acetylates p53/TP53 at 'Lys-120' and 'Lys-382' and controls its transcriptional activity via association with PML.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOZ
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Kat6a
Gene Accession No. Q92794
Gene Alias 1500036M03; 9930021N24Rik; histone acetyltransferase KAT6A; histone acetyltransferase MYST3; K(lysine) acetyltransferase 6A; KAT6A; lysine acetyltransferase 6A; monocytic leukemia zinc finger homolog; Monocytic leukemia zinc finger protein; Moz; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 3; MRD32; MYST histone acetyltransferase (monocytic leukemia) 3; Myst3; MYST-3; runt-related transcription factor binding protein 2; Runt-related transcription factor-binding protein 2; RUNXBP2; ZC2HC6A; Zfp220; zinc finger protein 220; ZNF220
Gene Symbols Kat6a
Host Species Rabbit
Immunogen Recombinant Human MOZ. Recombinant protein control fragment (Product #RP-107223).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7994
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.