Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ MOZ Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA571427
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA571427 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA571427 Supplier Invitrogen™ Supplier No. PA571427
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MOZ Polyclonal Antibody for Western Blot

Histone acetyltransferase that acetylates lysine residues in histone H3 and histone H4 (in vitro). Component of the MOZ/MORF complex which has a histone H3 acetyltransferase activity. May act as a transcriptional coactivator for RUNX1 and RUNX2. Acetylates p53/TP53 at 'Lys-120' and 'Lys-382' and controls its transcriptional activity via association with PML.
TRUSTED_SUSTAINABILITY

Specifications

Antigen MOZ
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Kat6a
Gene Accession No. Q92794
Gene Alias 1500036M03; 9930021N24Rik; histone acetyltransferase KAT6A; histone acetyltransferase MYST3; K(lysine) acetyltransferase 6A; KAT6A; lysine acetyltransferase 6A; monocytic leukemia zinc finger homolog; Monocytic leukemia zinc finger protein; Moz; MOZ, YBF2/SAS3, SAS2 and TIP60 protein 3; MRD32; MYST histone acetyltransferase (monocytic leukemia) 3; Myst3; MYST-3; runt-related transcription factor binding protein 2; Runt-related transcription factor-binding protein 2; RUNXBP2; ZC2HC6A; Zfp220; zinc finger protein 220; ZNF220
Gene Symbols Kat6a
Host Species Rabbit
Immunogen synthetic peptide directed towards the N terminal region of human MOZ.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7994
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.