Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Albumin, Bovine, Reagent Grade

Catalog No. p-135803
Click to view available options
Quantity:
50g

Prepared by heat shock fractionation

  • For IVD assays, immunoassays, enzyme stabilization
  • Low fatty acids, IgG
  • Purity: >98%
  • pH: 7

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1mg/mL
For Use With (Application) Immunoassay, Mammalian Cell Culture
Molecular Weight (g/mol) 66
Name Bovine Albumin Fraction V
pH Range 7
Quantity 50g
Storage Requirements 0° to 5°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.