Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

α-Amylase from Porcine pancreas, MP Biomedicals™

Catalog No. ICN19123950 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19123950 50 mg
ICN19123925 25 mg
MP219123910 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19123950 Supplier MP Biomedicals, Inc Supplier No. 0219123950
Add to Cart
Add to Cart

α-Amylase from Porcine pancreas ≥700 u/mg protein, Opaque, White Suspension

  • It is used to hydrolyze α bonds of α-linked polysaccharides, such as starch and glycogen.
  • It has been used in various plant studies, such as metabolism studies in Arabidopsis.
  • It is isolated from porcine pancreas is a glycoprotein.
  • It is a single polypeptide chain of ∽475 residues containing two SH groups and four disulfide bridges and a tightly bound Ca2+ necessary for stability. Chloride ions are necessary for activity and stability.
TRUSTED_SUSTAINABILITY

Specifications

Color Opaque White
pH 5.5 to 8.0 (activity range), 7 (optimum)
For Use With (Application) Molecular biology
Content And Storage 4°C
Source Porcine pancreas
Quantity 50 mg
Product Type alpha-Amylase
Synonym β-N-acetylglucosaminidase porcine placenta
Form Suspension
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.