Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Ascorbate Oxidase

Catalog No. ICN19007582 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kU
200 U
400 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19007582 200 U
ICN19007584 400 U
ICN19007591 1 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19007582 Supplier MP Biomedicals™ Supplier No. 0219007582
Add to Cart
Add to Cart

From Cucumis species

Activity: 200U/mg solid. Unit Definition: One unit will oxidize 1.0μmole of L-ascorbate to dehydroascorbate per minute at pH 5.6 and20 °C. Lyophilized powder.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥200 U/mg
Color Light Blue
pH 7 to 10
For Use With (Application) Enzymatic determination of Ascorbic Acid
Content And Storage 0°C
Source Cucumis species
Quantity 200 U
Product Type Ascorbate Oxidase
Molecular Weight (g/mol) 1320001, 140000
Inhibitors Azide, Cyanide, Na2S, Diethyldithiocarbamate (Na)
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.