Learn More
Description
Activity: ≥0.8U/mg solid. Unit Definition: One unit is the amount of enzyme that oxidizes one micromole of bilirubin per minute at 37°C, pH 8.4. Lyophilized powder.
Specifications
Specifications
| Concentration | ≥0.8 U/mg |
| Color | Light Blue/Green |
| For Use With (Application) | Degrade Bilirubin, used to determine free hemoglobin in icteric specimens, also has potential application in dye effluent decolorization and is a potential treatment for neonatal jaundice |
| Content And Storage | -20°C |
| Source | Myrothecium species |
| Quantity | 10 U |
| Product Type | Bilirubin Oxidase |
| Molecular Weight (g/mol) | 52000 |
| Form | Lyophilized Powder |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.