Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Albumin, from New Zealand-sourced Bovine Plasma

Catalog No. p-4949232
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
1 kg

Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies

  • From New Zealand-sourced Bovine Plasma
  • Microbiological Grade
  • ≥97% Total Protein
  • Yellow to Brown Powder
  • Synonyms: BSA, Bovine albumin, Albumin from bovine serum
  • CAS Number: 9048-46-8
  • EC Number: 232-936-2
  • MDL Number: MFCD00130384

TRUSTED_SUSTAINABILITY

Specifications

Formulation ≥97% Total Protein
Name Bovine Albumin
Quantity 25 g
Source Bovine
Form Powder
Purity or Quality Grade Microbiological Grade
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.