Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Bovine Fibrinogen
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5g
100g
Description
Prepared by extensive washing of cryoprecipitate followed by a buffer extraction to remove unwanted proteins and precipitation with ammonium sulfate. Lyophilized powder. Clottable protein >90%.
Physical Characteristics
- Appearance: Off-white powder
- Clottable Protein (anhydrous): ≥90%
- Moisture: ≤5%
- Reported: Total protein, pH
Specifications
Specifications
| For Use With (Application) | Mammalian Cell Culture |
| Name | Bovine Fibrinogen 90% Clottable from Bovine Plasma |
| pH Range | 6.4 |
| Quantity | 5g |
| Source | Bovine Plasma |
| Storage Requirements | <-10°C |
| Form | Lyophilized |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction