Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Bovine Transferrin, (HOLO)

Catalog No. p-4500875 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 mg
500 mg

Catalog No. ICN15233590

Supplier: MP Biomedicals™ 0215233590

Only null left
Add to Cart
Add to Cart

Suitable for regulating iron levels in cell culture

Transferrin is a 77kDa plasma glycoprotein with the capacity to bind ferric ions and as such is a source of bio-available iron. The product is frequently used as a nutrient in cell and microbial culture and as a media supplement in the production of bio-pharmaceutical products. Transferrin has also been shown to function as a factor in the growth stimulation of many cell types, such as granulocytes, macrophages and endothelial cells. Chromatographically purified.

Physical characteristics:

  • Appearance: Reddish-brown powder
  • Purity: ≥95% w/w
  • Iron: ≥1200 mg/g protein
  • Moisture: ≤5%
  • pH (3%): 6.0 to 8.0
  • Solubility: Dissolves in <20 min.
Microbial evlauation and control:
  • Bioburden: ≤100cfu/g
  • Endotoxin: ≤1 EU/mg
  • Mycoplasma: Not detected
  • Virus: Not detected
Certifications: ISO9000, cGMP, EDQM

TRUSTED_SUSTAINABILITY

Specifications

pH Range 6 to 8
Quantity 500 mg
Source Bovine Serum
Storage Requirements 4°C
Product Type Bovine Transferrin (HOLO)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.