Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Calmodulin, Purity: ≥99%
Click to view available options
:
1mg
Description
Activity: >13,000U/mg protein. Lyophilized with 1.7mM HEPES, pH 7, 30μM CaCl2. Unit Definition: One unit will activate 3',5'-cyclic-nucleotide phosphodiesterase to 50% Vmax. It may be used to activate CaM-dependent phosphodiesterase, calcineurin, CaM Kinases, etc. MW: 16.7kDa.

Specifications
Specifications
Concentration | 13000mg/mL |
Molecular Weight (g/mol) | 16.7 |
Name | Calmodulin from Bovine Brain |
pH Range | 7 |
Quantity | 1mg |
Source | Bovine Brain |
Storage Requirements | -20°C |
Form | Lyophilized |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction