Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CellMaxx™ Bovine Albumin, Low Endotoxin, Stem Cell Culture

Catalog No. p-4341552
Click to view available options
Quantity:
10 g
25 g
50 g
100 g
500 g
1000 g

Lipid-rich bovine serum albumin, suitable for stem cell culture and media formulation

Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells. Lipid-rich formulation can be substituted for Albumax™ II.

Physical characteristics:

  • Form: Off white to brown powder
  • Purity:≥98% (w/w)
  • Total Protein: ≥96%
  • pH (4%): 6.5 to 7.5
  • Solubility: Dissolves in 20 minutes at 20° to 25°C
  • Moisture: ≤5% w/w
Microbial evaluation and control:
  • Endotoxin: ≤1EU/mg
  • Mycoplasma: Negative
  • Bioburden: ≤100cfu/g
  • Virus (9CFR): Negative
  • Reported: Lipid content

Certifications: ISO9001, ISO13485, cGMP, EDQM

 

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Mammalian Cell Culture
Formulation Lipid-rich
Molecular Weight (g/mol) 66
Name CellMaxx™ Bovine Albumin, Low Endotoxin
pH Range 6.5 to 7.5
Purification Method Chromatography
Quantity 10 g
Source Bovine
Storage Requirements 4°C
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.