Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Cholesterol Oxidase (EC 1.1.3.6)

Catalog No. ICN19000650 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kU
200 U
50 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19000650 50 U
ICN19000682 200 U
ICN19000691 1 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19000650 Supplier MP Biomedicals™ Supplier No. 0219000650

Lyophilized powder

Activity: >1.5U/mg solid. Unit Definition: One unit is the amount of enzyme that will oxidize 1.0 micromole of cholesterol per minute at 37°C, pH 7.0.

TRUSTED_SUSTAINABILITY

Specifications

Concentration >1.5 U/mg
Color Dark Yellow
For Use With (Application) Used to determine serum Cholesterol
Content And Storage 0°C
Source Microorganism
Quantity 50 U
Product Type Cholesterol Oxidase
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.