Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Cholesterol Oxidase from Streptomyces species

Catalog No. ICN15067425 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 U
25 U
250 U
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15067425 25 U
ICN15067480 100 U
ICN15067483 250 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15067425 Supplier MP Biomedicals™ Supplier No. 0215067425
Add to Cart
Add to Cart

Cholesterol oxidase (CHOD) catalyzes the first step in cholesterol catabolism

CAS # : 9028-76-6

Form: Lyophilized powder

Activity: Approximately 15 units/mg solid

Stability: Stable at -20°C. A decrease in activity of approximately 10% may occur at 2 to 8°C within 6 months.

Stabilizers: Bovine serum albumin, sugars.

Isoelectric point: 5.1 ± 0.1 and 5.4 ± 0.1

Michaelis Constant: 4.3 x 10-5 M (Cholesterol)

Inhibitors: Ionic detergents, Hg++, Ag+

Optimum pH: 6.5 to 7.0

Optimum Temperature: 45° to 50°C

pH Stability: 5.0 to 10.0 (25°C, 20 hr)

Thermal Stability: below 45°C (pH 7.0, 15 minutes)

This enzyme is useful for the determination of cholesterol in serum by coupling with cholesterol esterase

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≈15 U/mg
Color Dark Yellow
pH 6.5 to 7
For Use With (Application) Catalyzes the first step in Cholesterol catabolism
Content And Storage 0°C
Source Streptomyces species
Quantity 25 U
Product Type Cholesterol Oxidase
Molecular Weight (g/mol) ≈34000
Inhibitors Ionic detergents, Hg++, Ag+
Form Lyophilized Powder
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.