Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Cod Liver Oil
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 qt.
4 qt.
Description
Cod liver oil is a nutritional supplement derived from liver of cod fish, it has high levels of the omega-3 fatty acids, EPA and DHA.
Specifications
Specifications
| Product Type | Animal Diet |
| Components | Vitamin A: min 850 USP μ/g, Vitamin D: min 85 USP μ/g |
| For Use With (Application) | Cod liver Oil is a source of Omega-3 Fatty Acid |
| Quantity | 1 qt. |
| Storage Requirements | 15°C to 30°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction