Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Collagen Type I from Calf Skin
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
10mg
100mg
250mg
Description
- CAS Number: 9007-34-5
- EC Number: 232-697-4
- MDL Number: MFCD00130825
- Format: Powder
- Soluble in 0.075M sodium citrate pH 3.7 and in dilute acetic acid
For attachment of primary cultures of epithelioid cells and many other cell types
Specifications
Specifications
| For Use With (Application) | Tumor Marker |
| pH Range | 3.7 |
| Quantity | 10mg |
| Source | Calf Skin |
| Product Type | Chorionic Gonadotropin from Human Pregnancy Urine |
| Content And Storage | -20°C |
| Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction