Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Collagenase, Type 4

Catalog No. ICN19511090 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19511090 500 mg
ICN19511025 25 mg
ICN19511080 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19511090 Supplier MP Biomedicals™ Supplier No. 0219511090
Add to Cart
Add to Cart

From Clostridium histolyticum

Pancreatic Islet Isolation Grade. Suggested for pancreatic islet isolation. Prepared to contain lower tryptic activity levels. Activity: ≥160U per mg solid. Unit Definition: One unit liberates peptides from collagen equivalent in ninhydrin color to 1.0μmol of L-leucine in 5 hr. at pH 7.4 and 37°C in presence of calcium ions. Dialyzed, lyophilized.

TRUSTED_SUSTAINABILITY

Specifications

Concentration >160 U/mg
Color Brown
For Use With (Application) For tissue dissociation combined with other Enzymes such as Elastase, Trypsin/Papain
Content And Storage 4°C
Source Clostridium histolyticum
Quantity 500 mg
Product Type Collagenase Type 4
Inhibitors Cysteine, EDTA or o-phenanthroline, alpha2-macroglobulin - a large plasma glycoprotein, alpha2-macroglobulin, alpha1-antitrypsin
Form Lyophilized Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.