Learn More
MP Biomedicals™ Collagenase Type II from Clostridium histolyticum
Shop All MP Biomedicals, Inc ProductsDescription
Collagenase degrades native helical collagen fibrils. It has an important role in connective tissue metabolism and is produced by specific cells involved in repairs and remodelling processes. It is a type II enzyme that contains greater clostripain activity.
- Specific activity:> 125 units/mg solid
- Unit definition: Liberates one μmole of L-leucine equivalents from collagen in 5 hours at 37°C, pH 7.5
- Presentation: brown shiny flakes
- Format: lyophilized powder
- pH: 7-9 for the A-a enzyme (optimum pH)
- Solubility: Soluble in water
- Pack size: 1g
Specifications
Specifications
| Concentration | 1 mg/mL |
| Color | Brown |
| pH | 7 to 9 |
| For Use With (Application) | Collagen structural and biosynthetic studies, release of cells from animal tissues |
| Storage Buffer | 1mg/mL of Enzyme in 0.05M TES with 0.36mM Calcium Chloride, pH 7.5 |
| Content And Storage | 4°C |
| Source | Clostridium histolyticum |
| Quantity | 1 g |
| Product Type | Collagenase Type II |
| Molecular Weight (g/mol) | 105000 and 57400 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.