Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ CSM-HIS-LEU-URA , Powdered Media

Catalog No. MP114531212 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP114531212 10 g
MP114531222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MP114531212 Supplier MP Biomedicals™ Supplier No. 114531212
Add to Cart
Add to Cart

Dropout supplement for S. cereviseae

  • Triple dropout
  • Complete supplement mixture of amino acids
  • Complete formulations
  • Dropout formulations

Formulation

  • Add 0.65g of CSM-His-Leu-Ura to each liter of DOB or DOBA and autoclave at 121°C for 25 minutes for complete synthetic defined triple dropout medium

Applications:

Yeast cell culture, Gene expression, Transcription regulation

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage 15°C to 30°C
Form Powder
Product Type Yeast Media
Formulation Adenine 10mg/L, L-Arginine HCl 50mg/L, L-Aspartic Acid 80mg/L, L-Histidine HCl 0mg/L, L-Isoleucine 50mg/L, L-Leucine 0mg/L, L-Lysine HCl 50mg/L, L-Methionine 20mg/L, L-Phenylalanine 50mg/L, L-Threonine 100mg/L, L-Tryptophan 50mg/L, L-Tyrosine 50mg/L, Uracil 0mg/L, L-Valine 140mg/L
For Use With (Application) For growth of Saccharomyces cerevisiae
Quantity 10 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.