Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Deoxyribonuclease I from Bovine pancreas, MP Biomedicals™

Catalog No. ICN19006210 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
0.5 mU
1 mU
10 mU
250 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19006210 10 mU
ICN10057583 250 mg
MP219006201 1 mU
MP21900625 0.5 mU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN19006210 Supplier MP Biomedicals, Inc Supplier No. 0219006210
Add to Cart
Add to Cart

Deoxyribonuclease I from Bovine pancreas 50,000 Dornase u/mg solid, White powder

  • Deoxyribonuclease from beef pancreas, DNase I, was first crystallized by Kunitz.
  • It is an endonuclease which splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide yielding 5'-phosphate terminated polynucleotides with a free hydroxyl group on position 3'.
  • The average chain of limit digest is a tetranucleotide. DNase I acts upon single chain DNA, and upon double-stranded DNA and chromatin.
  • In the latter case, although histones restrict susceptibility to nuclease action, over a period of time nearly all chromatin DNA is acted upon.
  • According to Mirsky and Silverman, this could result from the looseness of histone attachment to DNA.
  • They found that lysine-rich histones more effectively block DNase access to DNA than arginine-rich histones
TRUSTED_SUSTAINABILITY

Specifications

Color White
pH 7.8
For Use With (Application) Used as a genetic marker for forensic identification
Content And Storage 0°C
Source Bovine pancreas
Quantity 10 mU
Product Type Deoxyribonuclease I
Molecular Weight (g/mol) 31000
Inhibitors EDTA 7,8 and Sodium Dodecyl Sulfate
Synonym DNase I
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.