Learn More
Description
- Deoxyribonuclease from beef pancreas, DNase I, was first crystallized by Kunitz.
- It is an endonuclease which splits phosphodiester linkages, preferentially adjacent to a pyrimidine nucleotide yielding 5'-phosphate terminated polynucleotides with a free hydroxyl group on position 3'.
- The average chain of limit digest is a tetranucleotide. DNase I acts upon single chain DNA, and upon double-stranded DNA and chromatin.
- In the latter case, although histones restrict susceptibility to nuclease action, over a period of time nearly all chromatin DNA is acted upon.
- According to Mirsky and Silverman, this could result from the looseness of histone attachment to DNA.
- They found that lysine-rich histones more effectively block DNase access to DNA than arginine-rich histones
Specifications
Specifications
| Color | White |
| pH | 7.8 optimum (literature) |
| CAS | 9003-98-9 |
| Content And Storage | Store at -0°C. |
| MDL Number | MFCD00130918 |
| SMILES | * |
| IUPAC Name | Deoxyribonuclease |
| Molecular Weight (g/mol) | 0.00 |
| Quantity | 1 mU |
| Synonym | Deoxyribonucleate 5′-oligonucleotido-hydrolase |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.