Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Diastase

Catalog No. ICN10153925 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10153925 25 g
ICN10153980 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10153925 Supplier MP Biomedicals™ Supplier No. 0210153925
Add to Cart
Add to Cart

Malt

Diastase, [9000-92-4]

  • Activity: 1000 Lintner u/g
  • Melting Point: 175°C (dec)

TRUSTED_SUSTAINABILITY

Specifications

Concentration 1000 U/g
Color Off-White
For Use With (Application) Digests 50g of Starch in <30 min.
Content And Storage 4°C
Source Malt
Quantity 25 g
Product Type Diastase
Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.