Learn More
Description
- Use 26.7g powder per liter
- Contents/liter: 1.7g YNB, 5g Ammonium Sulfate, 20g Dextrose
Yeast cell culture, Gene expression
Specifications
Specifications
| Certifications/Compliance | Molecular biology Certified™ |
| Content And Storage | 15°C to 30°C |
| Form | Powder |
| Product Type | Yeast Media |
| Formulation | YNB 1.7g/L, Ammonium Sulfate 5g/L, Dextrose 20g/L |
| For Use With (Application) | For growth of Saccharomyces cerevisiae |
| Quantity | 454 g |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.