Learn More
MP Biomedicals™ Elastase 2X Crystallized, from Porcine Pancreas
Shop All MP Biomedicals, Inc ProductsDescription
Elastase was used to study human emphysema by induction of emphysema in rodents. It has been used in a study to investigate the design, synthesis and evaluation of biomimetic affinity ligands for elastases. Elastase from porcine pancreas has also been used in a study to investigate the purification and partial characterization of the pancreatic proteolytic enzymes trypsin, chymotrypsin, and elastase. It hydrolyzes peptide bonds, especially those adjacent to neutral amino acids.
Specifications
Specifications
| Concentration | 2X |
| Color | Milky |
| For Use With (Application) | Investigate the purification and partial characterization of the pancreatic proteolytic Enzymes |
| Content And Storage | 4°C |
| Source | Porcine pancreas |
| Quantity | 10 mg |
| Product Type | Elastase |
| Molecular Weight (g/mol) | ≈25000 |
| Inhibitors | DFP, Elastinal, alpha-2-macroglobulin |
| Form | Solution |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.