Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ FastPROTEIN™ Red Matrix

Catalog No. MP116550600
Click to view available options
Quantity:
100 Preps
50 Preps

Requires

FastPrep™ Instrument or alternative high speed disruptor, Microcentrifuge that can freely spin 2.0 mL tubes, Microcentrifuge tubes (2.0 mL and 1.5 mL)

FastPROTEIN™ Red Matrix Kit designed for the rapid extraction of proteins from yeast cells, fungi and algea in a few seconds.

  • Kit or Assay Type: Protein Isolation, Mini-prep
  • Molecule to be Enriched: Proteins
  • Sample Type: Yeast and Yeast Spores
  • Tube Size: 2 mL
  • Matrix Composition: Matrix C, 1.0 mm silica spheres
TRUSTED_SUSTAINABILITY

Specifications

Buffer Yeast breaking buffer
Product Type Red matrix kit
Quantity 50 Preps
Includes Matrix C, 1 mm Silica Spheres, Yeast breaking Buffer, Tubes
Content And Storage 15°C to 30°C
Capacity 2 mL
For Use With (Application) Rapid isolation and purification of Protein from Yeast cultures
Matrix FastProtein Red Matrix
WARNING: Cancer - www.P65Warnings.ca.gov

Unless specified otherwise, for research or further manufacturing use only, not for direct human use.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.