Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Geneclean™ Turbo Kit for PCR
Shop All MP Biomedicals, Inc Products
Click to view available options
No. of Reactions:
100 Preps
50 Preps
Description
- Ready for direct use
- Removes primers, nucleotides, enzymes, mineral oil, salts, and other impurities
- Applications: DNA Clean-up
Specifications
Specifications
| Sample Type | Post amplification PCR reaction mixture (oil-free) |
| For Use With (Application) | Isolation of DNA from PCR reactions, removal of primers, Nucleotides, Enzymes, Mineral Oil, salts and other impurities from DNA samples |
| Includes | GeneClean™ Turbo™ Salt Solution, Wash Solution, Cartridges, Catch Tubes and Elution Solution |
| No. of Reactions | 50 Preps |
| Sample Volume | Up to 100 μL PCR Reaction Solution |
| Sample Source | Gels and Solutions |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction