Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ γ-Globulin from Bovine Plasma

Catalog No. p-3573067 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
5 g
25 g
100 g

Purity: 98%

White, crystalline, lyophilized powder.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 25mg/mL
For Use With (Application) Cell Biology and Analysis, Genomics, Molecular Biology
Formulation The product is lyophilized, in normal Saline solution (25mg/mL, 0.9%NaCl)
Molecular Weight (g/mol) ∽150000
Name g-Globulin from Bovine Plasma
pH Range 6 to 8
Quantity 5 g
Source Bovine Plasma
Storage Requirements 0°C
Form Lyophilized
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.