Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ γ-Globulin, Bovine Purified

Catalog No. icn55847
Click to view available options
Quantity:
100 mg

Suitable for use as a carrier protein in secondary antibody systems, e.g., radioimmunoassay. Purified proteins are recommended as ELISA coating antigens and as controls.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 20 mg/mL
Quantity 100 mg
Content And Storage 2°C to 8°C
Form Lyophilized
For Use With (Application) Immunoassay
Product Type Purified Bovine gamma Globulin
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.