Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ γ-Globulins, Bovine Plasma, 100g
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 g
Specifications
Specifications
| Color | White to Off-white |
| Concentration | 100% |
| pH | 6.7 |
| Quantity | 100 g |
| Content And Storage | 2°C to 8°C |
| Form | Lyophilized |
| For Use With (Application) | Used as a co-precipitant in RIA methods employing Polyethylene Glycol or Ammonium Sulfate precipitate |
| Purity or Quality Grade | 99.3% |
| Host Species | Bovine Plasma |
| Product Type | Bovine Plasma gamma Globulin |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction