Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Glucose-6-Phosphate Dehydrogenase, From Yeast

Catalog No. ICN15118801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 kU
500 U
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15118801 1 kU
ICN1511885 500 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15118801 Supplier MP Biomedicals™ Supplier No. 0215118801

From Yeast

Glucose-6-Phosphate Dehydrogenase, [9001-40-5]

  • G-6-PDase
  • Salt-free
  • Lyophilized powder
  • Activity: -200 units/mg protein.
  • Unit Definition: One unit will cause the reduction of 1.0 ++mole of NADP+ per minute at pH 8.0 and 25°C.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≈200 U/mg
Color White
pH 5.5 to 7.5
For Use With (Application) Converts Glucose-6-phosphate into 6-phosphoglucono-d-lactone
Content And Storage -20°C
Source Yeast
Quantity 1 kU
Product Type Glucose-6-phosphate Dehydrogenase
Molecular Weight (g/mol) 104000
Inhibitors Acyl-CoA, Metal ions
Form Lyophilized Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.