Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ β-Glucosidase

Catalog No. ICN19519725 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 kU
25 kU
5 kU
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19519725 25 kU
ICN19519780 100 kU
ICN19519705 5 kU
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19519725 Supplier MP Biomedicals™ Supplier No. 0219519725

From Sweet Almonds

Activity: >1000U/mg material. Unit Definition: One unit is the amount of enzyme which liberates 1.0μg of glucose per minute at 35°C, pH 5.0. Salt-free. Lyophilized.

TRUSTED_SUSTAINABILITY

Specifications

Concentration ≥1000 U/mg
Color White
For Use With (Application) Synthesis of glucosides and fucosides
Content And Storage -20°C
Source Sweet almonds
Quantity 25 kU
Product Type beta-Glucosidase
Form Powder
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.