Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Gnome™ DNA Isolation Kit

Catalog No. p-4940230 Shop All MP Biomedicals, Inc Products
Click to view available options
No. of Reactions:
100 Preps
25 Preps

Catalog No. 11-201-0600

Supplier: MP Biomedicals™ 112010600

Only null left
Add to Cart
Add to Cart

Gnome™ DNA Isolation Kit offers a rapid and efficient method for isolation of high molecular weight genomic DNA from bacteria, plant, yeast, blood cells and other mammalian cells and tissues of all types.

  • No organic extractions are required, and up to 100 μg of DNA can be isolated per prep.
  • The GNOME™ kit utilizes RNase Mixx to eliminate RNA immediately after lysis, and Protease Mixx to rapidly degrade cellular proteins. This is followed by a proprietary ''salting-out'' technique which precludes the need for phenol, chloroform, or other organic extractions.
  • The DNA can either be spooled, removed with forceps, or centrifuged and dissolved in TE.
  • The GNOME™ DNA Isolation kit is used to quickly and efficiently isolate high molecular weight genomic DNA from cells and tissues of any type, including bacteria, yeast, plant, and animal cells.
  • Each preparation with the GNOME™ kit yields up to 100 μg of genomic DNA.
  • The DNA isolated by the GNOME™ procedure is suitable for restriction enzyme digestion or PCR amplification in as little as 1 hour after cell lysis.
TRUSTED_SUSTAINABILITY

Specifications

Format Kit
Sample Type Blood, Tissues, Yeast, Bacteria, Plant
Includes Cell Lysis/Denaturing Solution, RNase Mixx, Protease Mixx, Salt Out Mixture, Cell Suspension Solution
No. of Reactions 100 Preps
pH Cell Lysis/Denaturing Solution pH range: 6.2 to 6.9, Salt-out Mixture pH range: 6.0 to 6.8, Cell Suspension Solution pH range: 8.0 to 8.2
Product Type Gnome™ DNA Isolation Kit
Sample Volume 20 to 50 mg wet weight bacterial cell pellet, Up to 250 mg plant tissue, 2 to 5 x 107 nucleated white blood cells, 50 to 250 mg mammalian tissue, ∼1 to 5 x 108 yeast cell pellet
Yield 100 μg
WARNING: Cancer - www.P65Warnings.ca.gov

Research Use Only (RUO). Ready for CE IVD labeling of clinical applications.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.