Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ 1X Hanks' Balanced Salt Solution (Modified) without Phenol Red
Click to view available options
Quantity:
500 mL
Description
- Phenol red has been shown to interfere with the growth of some cells at cloning densities.
- Use this medium when working with stem cells or when growing cells at low densities.

Specifications
Specifications
Chemical Name or Material | Hanks' Balanced Salt Solution (HBSS) |
Color | Clear |
Concentration | 1 X |
pH | 6.9 to 7.4 |
Physical Form | Liquid |
Quantity | 500 mL |
Formulation | Calcium Chloride 140 mg/L, Magnesium Sulfate 97.7 mg/L, Potassium Chloride 400 mg/L, Potassium Phosphate Monobasic 60 mg/L, Sodium Bicarbonate 350 mg/L, Sodium Chloride 8000 mg/L, Sodium Phosphate Dibasic 47.7 mg/L, Dextrose 1000 mg/L |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction