Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Hemocyanin
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Description
Hemocyanins are copper-containing oxygen transport proteins found in the hemolymph of many invertebrates. It is dissolved in the hemolymph; not found in blood cells. They are divided into 2 main groups, arthropodan (such as horseshoe crab) and molluscan (such as keyhole limpet). These have structurally similar oxygen-binding sites, which are similar to the oxygen-binding sites of tyrosinases, but their quaternary structures are arranged differently.
- Format: Gray lyophilized powder
Immunology

Specifications
Specifications
Formulation | 5mM BES Buffer, pH 6.5 and 10mM MgSO4 |
Molecular Weight (g/mol) | 3000000 to 7500000 |
Name | Hemocyanin |
Quantity | 50 mg |
Source | Keyhole Limpet |
Storage Requirements | 4°C |
Form | Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction