Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ Human Growth Factors and Supplements
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
100 μg
Specifications
Specifications
| Content And Storage | -20°C |
| Product Type | Growth Factor |
| Format | Recombinant |
| Formulation | Sterile filtered lyophilized powder |
| For Use With (Application) | Supplementing cell cultures and investigating various metabolic processes |
| Purity or Quality Grade | >95% |
| Quantity | 100 μg |
| Reactivity | Human |
| Source | Recombinant, expressed in E. coli. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction