Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals™ IPTG Powder
Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
1 g
5 g
Description
Protein trafficking; Cell viability; Cytotoxicity; Gene expression; Transformation; Cloning and expression
Specifications
Specifications
| Content And Storage | -20°C |
| pH | 5 to 7 |
| Quantity | 5 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction