Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Lactoperoxidase, 60-80U/mg

Catalog No. ICN19527201 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
25 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19527201 1 mg
ICN19527210 10 mg
ICN19527205 5 mg
ICN19527225 25 mg
ICN19527280 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. ICN19527201 Supplier MP Biomedicals™ Supplier No. 0219527201

From bovine milk

Activity: 60-80U/mg protein. A412/A280 = 0.70 to 0.85. Unit Definition: One unit will form 1.0mg of purpurogallin from pyrogallol in 20 seconds at pH 6.0 and 20°C.

TRUSTED_SUSTAINABILITY

Specifications

Concentration 60 to 80 U/mg
Color Brown
For Use With (Application) Typically used for labelling Proteins with Radioiodine
Content And Storage 0°C
Source Bovine Milk
Quantity 1 mg
Product Type Lactoperoxidase
Molecular Weight (g/mol) 77500
Inhibitors Hydrazines, Cyanide
Form Lyophilized Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.