Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ LB-Agar, Powdered

Catalog No. MP113002275 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
454 g
1000 g
5000 g
10 x 500 mL
10 x 1000 mL
Format:
Capsule
Pouch
Powder
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
MP113002275 10 x 1000 mL Pouch
MP113002232 1000 g Powder
MP113002265 10 x 500 mL Pouch
MP113002222 454 g Powder
MP113002231 1000 g Capsule
MP113002241 5000 g Capsule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MP113002275 Supplier MP Biomedicals™ Supplier No. 113002275
Add to Cart
Add to Cart

Offered in a variety of formats for convenience and ease of use

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular biology Certified™
Content And Storage Room Temperature
Form Powder
Format Pouch
Product Type LB Agar
Formulation Tryptone 10g/L, Yeast extract 5g/L
For Use With (Application) For growth of bacteria
Packaging Type Pouch
Quantity 10 x 1000 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.